1-(3,4-Dichlorobenzyl)-4-piperidinecarbohydrazide ≥95%

Supplier: Matrix Scientific

045356-500MG 045356-1G 045356-5G
101870-472EA 318.46 USD
101870-472 101870-474 101870-476
1-(3,4-Dichlorobenzyl)-4-piperidinecarbohydrazide ≥95%
1-(3,4-Dichlorobenzyl)-4-piperidinecarbohydrazide

Hazard: Irritant

Formula: C₁₃H₁₇Cl₂N₃O
Melting Pt: 131…133 °C
MDL Number: MFCD03001279
CAS Number: 400878-29-7

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR