2-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045355-500MG 045355-1G 045355-5G
101870-466EA 461.54 USD
101870-466 101870-468 101870-470
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-thiophenecarboxylic acid ≥95%
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₂S
Melting Pt: 188…190 °C
MDL Number: MFCD01312401
CAS Number: 478077-98-4

Order Now

Specification Test Results

Melting point 188-190°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR