1-Methyl-1H-pyrrole-2-carbaldehyde oxime ≥95%

Supplier: Matrix Scientific

045351-500MG 045351-1G
101870-456EA 181.54 USD
101870-456 101870-458
1-Methyl-1H-pyrrole-2-carbaldehyde oxime ≥95%
1-Methyl-1H-pyrrole-2-carbaldehyde oxime

Hazard: Irritant

Formula: C₆H₈N₂O
Melting Pt: 146…148 °C
MDL Number: MFCD09054061
CAS Number: 37110-15-9

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR