1,1,1-Trifluoro-3-{[3-(trifluoromethyl)benzyl]amino}-2-propanol ≥95%

Supplier: Matrix Scientific

Synonyms: N-(2-Hydroxy-3,3,3-trifluoropropyl)-3-(trifluoromethyl)benzylamine

045350-500MG 045350-1G
101870-452EA 1109.23 USD
101870-452 101870-454
1,1,1-Trifluoro-3-{[3-(trifluoromethyl)benzyl]amino}-2-propanol ≥95%
1,1,1-Trifluoro-3-{[3-(trifluoromethyl)benzyl]amino}-2-propanol

Hazard: Irritant

Formula: C₁₁H₁₁F₆NO
MW: 287.21 g/mol
Melting Pt: 65…67 °C
MDL Number: MFCD01871640
CAS Number: 478068-12-1

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR