3-Amino-3-[4-(trifluoromethyl)phenyl]acrylonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: (2Z)-3-Amino-3-[4-(trifluoromethyl)phenyl]prop-2-enenitrile

045349-500MG 045349-1G 045349-5G
101870-446EA 181.54 USD
101870-446 101870-448 101870-450
3-Amino-3-[4-(trifluoromethyl)phenyl]acrylonitrile ≥95%
3-Amino-3-[4-(trifluoromethyl)phenyl]acrylonitrile

Hazard: Irritant

Formula: C₁₀H₇F₃N₂
MW: 212.18 g/mol
Melting Pt: 137…139 °C
MDL Number: MFCD01871638
CAS Number: 71682-89-8

Order Now

Specification Test Results

Melting point 137-139°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR