3-Amino-3-(4-bromophenyl)acrylonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: (2E)-3-Amino-3-(4-bromophenyl)prop-2-enenitrile

045348-500MG 045348-1G 045348-5G
101870-440EA 181.54 USD
101870-440 101870-442 101870-444
3-Amino-3-(4-bromophenyl)acrylonitrile ≥95%
3-Amino-3-(4-bromophenyl)acrylonitrile

Hazard: Irritant

Formula: C₉H₇BrN₂
MW: 223.08 g/mol
Melting Pt: 148…150 °C
MDL Number: MFCD01871637
CAS Number: 748807-82-1

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR