1,1,1-Trifluoro-3-(phenethylamino)-2-propanol ≥95%

Supplier: Matrix Scientific

045345-500MG 045345-1G 045345-5G
101870-434EA 673.85 USD
101870-434 101870-436 101870-438
1,1,1-Trifluoro-3-(phenethylamino)-2-propanol ≥95%
1,1,1-Trifluoro-3-(phenethylamino)-2-propanol

Hazard: Irritant

Formula: C₁₁H₁₄F₃NO
Melting Pt: 63-65°C
MDL Number: MFCD03001278
CAS Number: 400878-20-8

Order Now

Specification Test Results

Melting point 63-65°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR