2-(4-Bromophenyl)-1,3-thiazolane-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

045344-500MG 045344-1G 045344-5G
101870-428EA 80 USD
101870-428 101870-430 101870-432
2-(4-Bromophenyl)-1,3-thiazolane-4-carboxylic acid ≥95%
2-(4-Bromophenyl)-1,3-thiazolane-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₀BrNO₂S
Melting Pt: 175…177 °C
MDL Number: MFCD01067251
CAS Number: 69570-83-8

Order Now

Specification Test Results

Melting point 175-177°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR