2-[(3,4,4-Trifluoro-3-butenyl)sulfanyl]nicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(3,4,4-Trifluorobut-3-en-1-yl)thio]nicotinic acid

045343-500MG 045343-1G 045343-5G
101870-422EA 375.38 USD
101870-422 101870-424 101870-426
2-[(3,4,4-Trifluoro-3-butenyl)sulfanyl]nicotinic acid ≥95%
2-[(3,4,4-Trifluoro-3-butenyl)sulfanyl]nicotinic acid

Hazard: Irritant

Formula: C₁₀H₈F₃NO₂S
MW: 263.24 g/mol
Melting Pt: 123…125 °C
MDL Number: MFCD01814935
CAS Number: 453557-66-9

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR