2-(4-Iodophenoxy)pyrimidine ≥95%

Supplier: Matrix Scientific

045336-500MG 045336-1G 045336-5G
101870-380EA 181.54 USD
101870-380 101870-382 101870-384
2-(4-Iodophenoxy)pyrimidine ≥95%
2-(4-Iodophenoxy)pyrimidine

Hazard: Irritant

Formula: C₁₀H₇IN₂O
Melting Pt: 87…90 °C
MDL Number: MFCD01443687
CAS Number: 330792-86-4

Order Now

Specification Test Results

Melting point 87-90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR