4-(2-Pyrimidinyloxy)aniline ≥95%

Supplier: Matrix Scientific

045335-500MG 045335-1G 045335-5G
101870-374EA 278.46 USD
101870-374 101870-376 101870-378
4-(2-Pyrimidinyloxy)aniline ≥95%
4-(2-Pyrimidinyloxy)aniline

Hazard: Irritant

Formula: C₁₀H₉N₃O
Melting Pt: 140…142 °C
MDL Number: MFCD01443685
CAS Number: 105130-26-5

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR