2-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-1H-isoindole-1,3(2H)-dione ≥95%
Supplier: Matrix Scientific
045334-500MG
045334-1G
045334-5G
101870-368EA
181.54
USD
101870-368
101870-370
101870-372
2-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-1H-isoindole-1,3(2H)-dione ≥95%
2-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-1H-isoindole-1,3(2H)-dione
Hazard: Irritant
Formula:
C₁₂H₇ClN₂O₃S Melting Pt: 152…154 °C |
MDL Number:
MFCD01443862 CAS Number: 339018-42-7 |
Specification Test Results
Melting point | 152-154°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...