N-(4-Methoxybenzyl)urea ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-Methoxybenzyl)urea
045330-500MG
045330-1G
045330-5G
101870-344EA
181.54
USD
101870-344
101870-346
101870-348
N-(4-Methoxybenzyl)urea ≥95%
N-(4-Methoxybenzyl)urea
Hazard: Irritant
Formula:
C₉H₁₂N₂O₂ MW: 180.21 g/mol Melting Pt: 160…162 °C |
MDL Number:
MFCD01314623 CAS Number: 54582-35-3 |
Specification Test Results
Melting point | 160-162°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...