4,5-Dibromo-3(2H)-pyridazinone ≥95%
Supplier: Matrix Scientific
Synonyms:
4,5-Dibromopyridazin-3(2H)-one
045328-5G
045328-25G
101870-336EA
153.85
USD
101870-336
102833-134
4,5-Dibromo-3(2H)-pyridazinone ≥95%
4,5-Dibromo-3(2H)-pyridazinone
Hazard: Irritant
Formula:
C₄H₂Br₂N₂O MW: 253.89 g/mol Melting Pt: 231-233°C |
MDL Number:
MFCD00023641 CAS Number: 5788-58-9 |
Specification Test Results
Melting point | 231-233°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...