1-(4-Bromophenyl)-3-(dimethylamino)-2-buten-1-one ≥97%

Supplier: Matrix Scientific

045327-500MG 045327-1G 045327-5G
101870-326EA 278.46 USD
101870-326 101870-328 101870-330
1-(4-Bromophenyl)-3-(dimethylamino)-2-buten-1-one ≥97%
1-(4-Bromophenyl)-3-(dimethylamino)-2-buten-1-one

Hazard: Irritant

Formula: C₁₂H₁₄BrNO
MW: 268.16 g/mol
Melting Pt: 115…116 °C
MDL Number: MFCD08056661
CAS Number: 178261-64-8

Order Now

Specification Test Results

Melting point 115-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR