2-Bromo-1-(6-methyl[1,3]thiazolo[3,2-b] [1,2,4]triazolo-5-yl)-1-ethanone hydrobromide ≥95%

Supplier: Matrix Scientific

045326-500MG 045326-1G 045326-5G
101870-320EA 747.69 USD
101870-320 101870-322 101870-324
2-Bromo-1-(6-methyl[1,3]thiazolo[3,2-b] [1,2,4]triazolo-5-yl)-1-ethanone hydrobromide ≥95%
2-Bromo-1-(6-methyl[1,3]thiazolo[3,2-b] [1,2,4]triazolo-5-yl)-1-ethanone hydrobromide

Hazard: Irritant

Formula: C₇H₇Br₂N₃OS
Melting Pt: 218 °C
MDL Number: MFCD03001276
CAS Number: 187597-19-9

Order Now

Specification Test Results

Melting point 218°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR