3-(Dimethylamino)-1-phenyl-2-buten-1-one ≥95%
Supplier: Matrix Scientific
045324-500MG
045324-1G
045324-5G
101870-308EA
278.46
USD
101870-308
101870-310
101870-312
3-(Dimethylamino)-1-phenyl-2-buten-1-one ≥95%
3-(Dimethylamino)-1-phenyl-2-buten-1-one
Hazard: Irritant
Formula:
C₁₂H₁₅NO Melting Pt: 68…70 °C |
MDL Number:
MFCD08056660 CAS Number: 34523-87-0 |
Specification Test Results
Melting point | 68-70°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...