2-[1-(3-Chlorobenzyl)-1H-indol-3-yl]acetic acid ≥95%

Supplier: Matrix Scientific

045317-500MG 045317-1G 045317-5G
101870-274EA 278.46 USD
101870-274 101870-276 101870-278
2-[1-(3-Chlorobenzyl)-1H-indol-3-yl]acetic acid ≥95%
2-[1-(3-Chlorobenzyl)-1H-indol-3-yl]acetic acid

Hazard: Irritant

Formula: C₁₇H₁₄ClNO₂
MW: 299.76 g/mol
Melting Pt: 105…107 °C
MDL Number: MFCD00794689
CAS Number: 339016-33-0

Order Now

Specification Test Results

Melting point 105-107°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR