2-[1-(6-Chloro-3-pyridazinyl)-1H-indol-3-yl]acetic acid ≥95%

Supplier: Matrix Scientific

045316-500MG 045316-1G 045316-5G
101870-268EA 375.38 USD
101870-268 101870-270 101870-272
2-[1-(6-Chloro-3-pyridazinyl)-1H-indol-3-yl]acetic acid ≥95%
2-[1-(6-Chloro-3-pyridazinyl)-1H-indol-3-yl]acetic acid

Hazard: Irritant

Formula: C₁₄H₁₀ClN₃O₂
Melting Pt: 220…222 °C
MDL Number: MFCD00794687
CAS Number: 339016-30-7

Order Now

Specification Test Results

Melting point 220-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR