2-[1-(4-Fluorobenzyl)-1H-indol-3-yl]acetic acid ≥95%

Supplier: Matrix Scientific

045315-500MG 045315-1G 045315-5G
101870-262EA 278.46 USD
101870-262 101870-264 101870-266
2-[1-(4-Fluorobenzyl)-1H-indol-3-yl]acetic acid ≥95%
2-[1-(4-Fluorobenzyl)-1H-indol-3-yl]acetic acid

Hazard: Irritant

Formula: C₁₇H₁₄FNO₂
Melting Pt: 163…166 °C
MDL Number: MFCD00794686
CAS Number: 176204-51-6

Order Now

Specification Test Results

Melting point 163-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR