N-(4-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide ≥95%

Supplier: Matrix Scientific

045313-500MG 045313-1G 045313-5G
101870-250EA 181.54 USD
101870-250 101870-252 101870-254
N-(4-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide ≥95%
N-(4-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₃H₁₆ClNO₂
Melting Pt: 105…107 °C
MDL Number: MFCD00793413
CAS Number: 339015-92-8

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR