5-Amino-2,4-dichlorophenyl ethyl carbonate ≥95%

Supplier: Matrix Scientific

045311-500MG 045311-1G 045311-5G
101870-244EA 375.38 USD
101870-244 101870-246 101870-248
5-Amino-2,4-dichlorophenyl ethyl carbonate ≥95%
5-Amino-2,4-dichlorophenyl ethyl carbonate

Hazard: Irritant

Formula: C₉H₉Cl₂NO₃
Melting Pt: 58…60 °C
MDL Number: MFCD00974509
CAS Number: 339014-89-0

Order Now

Specification Test Results

Melting point 58-60°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR