4-Chloro-N-(5-chloro-2-pyridinyl)butanamide ≥95%

Supplier: Matrix Scientific

045309-500MG 045309-1G 045309-5G
101870-232EA 181.54 USD
101870-232 101870-234 101870-236
4-Chloro-N-(5-chloro-2-pyridinyl)butanamide ≥95%
4-Chloro-N-(5-chloro-2-pyridinyl)butanamide

Hazard: Irritant

Formula: C₉H₁₀Cl₂N₂O
MW: 233.1 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD00664609
CAS Number: 339014-78-7

Order Now

Specification Test Results

Melting point 110-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR