2-(1H-Pyrrol-1-yl)benzenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1H-Pyrrol-1-yl)phenol

045307-500MG 045307-1G 045307-5G
101870-220EA 278.46 USD
101870-220 101870-222 101870-224
2-(1H-Pyrrol-1-yl)benzenol ≥95%
2-(1H-Pyrrol-1-yl)benzenol

Hazard: Irritant

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Melting Pt: 48…50 °C
MDL Number: MFCD00128327
CAS Number: 32277-91-1

Order Now

Specification Test Results

Melting point 48-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR