2-[5-(2,4-Dichlorobenzoyl)-2-thienyl]acetonitrile ≥95%

Supplier: Matrix Scientific

045306-500MG 045306-1G 045306-5G
101870-214EA 278.46 USD
101870-214 101870-216 101870-218
2-[5-(2,4-Dichlorobenzoyl)-2-thienyl]acetonitrile ≥95%
2-[5-(2,4-Dichlorobenzoyl)-2-thienyl]acetonitrile

Hazard: Irritant

Formula: C₁₃H₇Cl₂NOS
Melting Pt: 130…132 °C
MDL Number: MFCD00664601
CAS Number: 339014-61-8

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR