O-(4-Nitrobenzyl)hydroxylamine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
O-(4-Nitrobenzyl)-hydroxylammonium chloride
045305-500MG
045305-1G
045305-5G
101870-208EA
40
USD
101870-208
101870-210
101870-212
O-(4-Nitrobenzyl)hydroxylamine hydrochloride ≥95%
O-(4-Nitrobenzyl)hydroxylamine hydrochloride
Hazard: Irritant
Formula:
C₇H₉ClN₂O₃ MW: 204.61 g/mol Melting Pt: 226°C |
MDL Number:
MFCD00012954 CAS Number: 2086-26-2 |
Specification Test Results
Melting point | 226°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...