2-Amino-1-(2-naphthyl)-1-ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Amino-1-hydroxyethyl)naphthalene

045303-500MG 045303-1G 045303-5G
101870-202EA 453.85 USD
101870-202 101870-204 101870-206
2-Amino-1-(2-naphthyl)-1-ethanol ≥95%
2-Amino-1-(2-naphthyl)-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₃NO
MW: 187.24 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00974251
CAS Number: 5696-74-2

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR