2-Amino-1-(2-naphthyl)-1-ethanone hydrochloride ≥95%

Supplier: Matrix Scientific

045302-500MG 045302-1G 045302-5G
101870-196EA 375.38 USD
101870-196 101870-198 101870-200
2-Amino-1-(2-naphthyl)-1-ethanone hydrochloride ≥95%
2-Amino-1-(2-naphthyl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₂ClNO
Melting Pt: 226…228 °C
MDL Number: MFCD08056659
CAS Number: 38061-36-8

Order Now

Specification Test Results

Melting point 226-228°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR