(4-Methoxycarbonylbenzyl)triphenylphosphonium bromide ≥95%

Supplier: Matrix Scientific

Synonyms: [4-(Methoxycarbonyl)benzyl](triphenyl)phosphonium bromide

045301-500MG 045301-1G 045301-5G
101870-190EA 375.38 USD
101870-190 101870-192 101870-194
(4-Methoxycarbonylbenzyl)triphenylphosphonium bromide ≥95%
(4-Methoxycarbonylbenzyl)triphenylphosphonium bromide

Hazard: Irritant

Formula: C₂₇H₂₄BrO₂P
Melting Pt: 255…257 °C
MDL Number: MFCD00082765
CAS Number: 1253-46-9

Order Now

Specification Test Results

Melting point 255-257°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR