2-(Bromoacetyl)naphthalene ≥95%

Supplier: Matrix Scientific

Synonyms: α-Bromo-2'-acetonaphthone, 2-Bromo-2'-acetonaphthone

045300-25G 045300-100G
101870-186EA 200 USD
101870-186 101870-188
2-(Bromoacetyl)naphthalene ≥95%
2-(Bromoacetyl)naphthalene

Hazard: Irritant

Formula: C₁₂H₉BrO
MW: 249.11 g/mol
Melting Pt: 86-88°C
MDL Number: MFCD00004109
CAS Number: 613-54-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR