2-(Benzylsulfanyl)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
045299-500MG
045299-1G
045299-5G
101870-180EA
221.54
USD
101870-180
101870-182
101870-184
2-(Benzylsulfanyl)benzenecarboxylic acid ≥95%
2-(Benzylsulfanyl)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₂O₂S Melting Pt: 180…182 °C |
MDL Number:
MFCD00995231 CAS Number: 1531-80-2 |
Specification Test Results
Melting point | 180-182°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...