6-Methyl-3-(4-methylphenyl)-1,2,4-triazin-5-ol ≥95%

Supplier: Matrix Scientific

045295-500MG 045295-1G 045295-5G
101870-158EA 375.38 USD
101870-158 101870-160 101870-162
6-Methyl-3-(4-methylphenyl)-1,2,4-triazin-5-ol ≥95%
6-Methyl-3-(4-methylphenyl)-1,2,4-triazin-5-ol

Hazard: Irritant

Formula: C₁₁H₁₁N₃O
MW: 201.23 g/mol
Melting Pt: 247…248 °C
MDL Number: MFCD04117816
CAS Number: 109306-98-1

Order Now

Specification Test Results

Melting point 247-248°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR