5-Chloro-3,6-diphenyl-1,2,4-triazine ≥95%

Supplier: Matrix Scientific

045293-500MG 045293-1G 045293-5G
101870-146EA 556.92 USD
101870-146 101870-148 101870-150
5-Chloro-3,6-diphenyl-1,2,4-triazine ≥95%
5-Chloro-3,6-diphenyl-1,2,4-triazine

Hazard: Irritant

Formula: C₁₅H₁₀ClN₃
MW: 267.72 g/mol
Melting Pt: 132…134 °C
MDL Number: MFCD08056658
CAS Number: 94398-27-3

Order Now

Specification Test Results

Melting point 132-134°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR