3,6-Diphenyl-1,2,4-triazin-5-ol ≥95%

Supplier: Matrix Scientific

045292-500MG 045292-1G 045292-5G
101870-140EA 375.38 USD
101870-140 101870-142 101870-144
3,6-Diphenyl-1,2,4-triazin-5-ol ≥95%
3,6-Diphenyl-1,2,4-triazin-5-ol

Hazard: Irritant

Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Melting Pt: 264…266 °C
MDL Number: MFCD07755895
CAS Number: 36214-25-2

Order Now

Specification Test Results

Melting point 264-266°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR