2-[Cyano(3,4-dimethoxyphenyl)methyl]-6-fluorobenzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Cyano-3-fluorophenyl)-2-(3,4-dimethoxyphenyl)acetonitrile

045291-500MG 045291-1G 045291-5G
101870-134EA 181.54 USD
101870-134 101870-136 101870-138
2-[Cyano(3,4-dimethoxyphenyl)methyl]-6-fluorobenzenecarbonitrile ≥95%
2-[Cyano(3,4-dimethoxyphenyl)methyl]-6-fluorobenzenecarbonitrile

Hazard: Irritant

Formula: C₁₇H₁₃FN₂O₂
MW: 296.3 g/mol
Melting Pt: 160…161 °C
MDL Number: MFCD00231998
CAS Number: 339012-94-1

Order Now

Specification Test Results

Melting point 160-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR