5-(3-Chloro-2-thienyl)-1H-pyrazole ≥95%
Supplier: Matrix Scientific
045289-500MG
045289-1G
045289-5G
101870-124EA
747.69
USD
101870-124
101870-126
101870-128
5-(3-Chloro-2-thienyl)-1H-pyrazole ≥95%
5-(3-Chloro-2-thienyl)-1H-pyrazole
Hazard: Irritant
Formula:
C₇H₅ClN₂S MW: 184.65 g/mol Melting Pt: 112…114 °C |
MDL Number:
MFCD00793877 CAS Number: 166196-59-4 |
Specification Test Results
Melting point | 112-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...