4-(4-Chlorophenyl)-2-(4-isopropylphenyl)-4-oxobutanoic acid ≥95%

Supplier: Matrix Scientific

045285-500MG 045285-1G
101870-106EA 556.92 USD
101870-106 101870-108
4-(4-Chlorophenyl)-2-(4-isopropylphenyl)-4-oxobutanoic acid ≥95%
4-(4-Chlorophenyl)-2-(4-isopropylphenyl)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₉H₁₉ClO₃
Melting Pt: 154…157 °C
MDL Number: MFCD00664417
CAS Number: 344281-36-3

Order Now

Specification Test Results

Melting point 154-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR