2-(3-Chlorophenyl)-4-(4-chlorophenyl)-4-oxobutanoic acid ≥95%

Supplier: Matrix Scientific

045282-500MG 045282-1G
101870-094EA 556.92 USD
101870-094 101870-096
2-(3-Chlorophenyl)-4-(4-chlorophenyl)-4-oxobutanoic acid ≥95%
2-(3-Chlorophenyl)-4-(4-chlorophenyl)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₆H₁₂Cl₂O₃
Melting Pt: 179…182 °C
MDL Number: MFCD00664385
CAS Number: 344280-32-6

Order Now

Specification Test Results

Melting point 179-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR