2-(3-Chlorophenyl)-4-oxo-4-phenylbutanoic acid ≥95%

Supplier: Matrix Scientific

045280-500MG 045280-1G
101870-086EA 556.92 USD
101870-086 101870-088
2-(3-Chlorophenyl)-4-oxo-4-phenylbutanoic acid ≥95%
2-(3-Chlorophenyl)-4-oxo-4-phenylbutanoic acid

Hazard: Irritant

Formula: C₁₆H₁₃ClO₃
Melting Pt: 159…161 °C
MDL Number: MFCD00664378
CAS Number: 344280-12-2

Order Now

Specification Test Results

Melting point 159-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR