2,3-Dichloro-4-(ethylsulfonyl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045279-500MG 045279-1G 045279-5G
101870-080EA 460 USD
101870-080 101870-082 101870-084
2,3-Dichloro-4-(ethylsulfonyl)benzenecarboxylic acid ≥95%
2,3-Dichloro-4-(ethylsulfonyl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₉H₈Cl₂O₄S
Melting Pt: 183…186 °C
MDL Number: MFCD01935963
CAS Number: 105917-76-8

Order Now

Specification Test Results

Melting point 183-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR