1-[2,3-Dichloro-4-(ethylsulfonyl)phenyl]-1-ethanone ≥95%

Supplier: Matrix Scientific

045277-500MG 045277-1G 045277-5G
101870-074EA 747.69 USD
101870-074 101870-076 101870-078
1-[2,3-Dichloro-4-(ethylsulfonyl)phenyl]-1-ethanone ≥95%
1-[2,3-Dichloro-4-(ethylsulfonyl)phenyl]-1-ethanone

Hazard: Irritant

Formula: C₁₀H₁₀Cl₂O₃S
MW: 281.16 g/mol
Melting Pt: 88…90 °C
MDL Number: MFCD01935964
CAS Number: 338982-46-0

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR