1-[2,3-Dichloro-4-(ethylsulfanyl)phenyl]-1-ethanone ≥95%

Supplier: Matrix Scientific

045276-500MG 045276-1G 045276-5G
101870-068EA 375.38 USD
101870-068 101870-070 101870-072
1-[2,3-Dichloro-4-(ethylsulfanyl)phenyl]-1-ethanone ≥95%
1-[2,3-Dichloro-4-(ethylsulfanyl)phenyl]-1-ethanone

Hazard: Irritant

Formula: C₁₀H₁₀Cl₂OS
Melting Pt: 55…56 °C
MDL Number: MFCD05663800
CAS Number: 105917-69-9

Order Now

Specification Test Results

Melting point 55-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR