2-(4-Fluorophenoxy)ethanimidamide hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Fluorophenoxy)acetamidine hydrochloride

045274-500MG 045274-1G 045274-5G
101870-062EA 375.38 USD
101870-062 101870-064 101870-066
2-(4-Fluorophenoxy)ethanimidamide hydrochloride ≥95%
2-(4-Fluorophenoxy)ethanimidamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₀ClFN₂O
MW: 204.63 g/mol
Melting Pt: 156…159 °C
MDL Number: MFCD01114620
CAS Number: 318951-78-9

Order Now

Specification Test Results

Melting point 156-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR