3-[3-(4-Methylphenyl)-2,1-benzisoxazol-5-yl]acrylic acid ≥95%

Supplier: Matrix Scientific

045272-500MG 045272-1G 045272-5G
101870-050EA 1109.23 USD
101870-050 101870-052 101870-054
3-[3-(4-Methylphenyl)-2,1-benzisoxazol-5-yl]acrylic acid ≥95%
3-[3-(4-Methylphenyl)-2,1-benzisoxazol-5-yl]acrylic acid

Hazard: Irritant

Formula: C₁₇H₁₃NO₃
Melting Pt: 295 °C
MDL Number: MFCD00663847
CAS Number: 344276-77-3

Order Now

Specification Test Results

Melting point 295°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR