3-Phenyl-2,1-benzisoxazole-5-carbaldehyde ≥95%
Supplier: Matrix Scientific
045269-500MG
045269-1G
045269-5G
101870-032EA
376.92
USD
101870-032
101870-034
101870-036
3-Phenyl-2,1-benzisoxazole-5-carbaldehyde ≥95%
3-Phenyl-2,1-benzisoxazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₁₄H₉NO₂ Melting Pt: 158-160°C |
MDL Number:
MFCD00623237 CAS Number: 94752-06-4 |
Specification Test Results
Melting point | 158-160°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...