1-(3,4-Dichlorophenyl)-2-(methylsulfonyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

045268-500MG 045268-1G 045268-5G
101870-026EA 278.46 USD
101870-026 101870-028 101870-030
1-(3,4-Dichlorophenyl)-2-(methylsulfonyl)-1-ethanone ≥95%
1-(3,4-Dichlorophenyl)-2-(methylsulfonyl)-1-ethanone

Hazard: Irritant

Formula: C₉H₈Cl₂O₃S
MW: 267.13 g/mol
Melting Pt: 125…126 °C
MDL Number: MFCD00202208
CAS Number: 344275-49-6

Order Now

Specification Test Results

Melting point 125-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR