5-{[(2,4-Dichlorobenzyl)sulfanyl]methyl}-4-isopropyl-4H-1,2,4-triazole-3-thiol ≥95%

Supplier: Matrix Scientific

045267-500MG 045267-1G 045267-5G
101870-020EA 747.69 USD
101870-020 101870-022 101870-024
5-{[(2,4-Dichlorobenzyl)sulfanyl]methyl}-4-isopropyl-4H-1,2,4-triazole-3-thiol ≥95%
5-{[(2,4-Dichlorobenzyl)sulfanyl]methyl}-4-isopropyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₃H₁₅Cl₂N₃S₂
Melting Pt: 137…140 °C
MDL Number: MFCD00244456
CAS Number: 344272-66-8

Order Now

Specification Test Results

Melting point 137-140°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR