5-{[(4-Chlorobenzyl)sulfanyl]methyl}-4-ethyl-4H-1,2,4-triazol-3-ylhydrosulfide ≥95%

Supplier: Matrix Scientific

045265-500MG 045265-1G 045265-5G
101870-008EA 747.69 USD
101870-008 101870-010 101870-012
5-{[(4-Chlorobenzyl)sulfanyl]methyl}-4-ethyl-4H-1,2,4-triazol-3-ylhydrosulfide ≥95%
5-{[(4-Chlorobenzyl)sulfanyl]methyl}-4-ethyl-4H-1,2,4-triazol-3-ylhydrosulfide

Hazard: Irritant

Formula: C₁₂H₁₄ClN₃S₂
Melting Pt: 148…150 °C
MDL Number: MFCD00244393
CAS Number: 344268-56-0

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR