5-{[(4-Chlorobenzyl)sulfanyl]methyl}-4-isopropyl-4H-1,2,4-triazol-3-ylhydrosulfide ≥95%

Supplier: Matrix Scientific

045264-500MG 045264-1G 045264-5G
101870-002EA 747.69 USD
101870-002 101870-004 101870-006
5-{[(4-Chlorobenzyl)sulfanyl]methyl}-4-isopropyl-4H-1,2,4-triazol-3-ylhydrosulfide ≥95%
5-{[(4-Chlorobenzyl)sulfanyl]methyl}-4-isopropyl-4H-1,2,4-triazol-3-ylhydrosulfide

Hazard: Irritant

Formula: C₁₃H₁₆ClN₃S₂
Melting Pt: 132…135 °C
MDL Number: MFCD01935945
CAS Number: 338982-45-9

Order Now

Specification Test Results

Melting point 132-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR