1-(4-Methoxyphenyl)-2-(methylsulfonyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

045263-500MG 045263-1G 045263-5G
101869-996EA 278.46 USD
101869-996 101869-998 101870-000
1-(4-Methoxyphenyl)-2-(methylsulfonyl)-1-ethanone ≥95%
1-(4-Methoxyphenyl)-2-(methylsulfonyl)-1-ethanone

Hazard: Irritant

Formula: C₁₀H₁₂O₄S
MW: 228.27 g/mol
Melting Pt: 136…137 °C
MDL Number: MFCD00202206
CAS Number: 27918-36-1

Order Now

Specification Test Results

Melting point 136-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR